missing translation for 'onlineSavingsMsg'
Learn More

Human DC-SIGN/CD209 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  R&D Systems FAB161G-100UG

Product Code. 18766618

  • ONLINE EXCLUSIVE PRICE
    € 563.00
    € 532.35 / 100µg
    Save € 30.65
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DC-SIGN/CD209
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Mouse
100 μg
Primary
Detects human DC-SIGN/CD209 on transfected NIH/3T3 cells and on monocyte derived dendritic cells. Does not react with parental mouse cells or irrelevant transfectants, such as human DC-SIGN2.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Flow Cytometry
120507
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD209
NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGN/CD209
RUO
30835
Human
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only (RUO)