missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DEC-205/CD205 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2047R-100UG
This item is not returnable.
View return policy
Description
DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Human samples. It is validated for Flow Cytometry.
Specifications
| DEC-205/CD205 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O60449 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 523203 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| LY75 | |
| E. coli-derived recombinant human DEC-205/CD205 Cys216-Cys501 Accession # O60449 | |
| RUO | |
| Detects human DEC-205/CD205 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse DEC-205 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction