missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DEFB4B (O15263, 24 a.a. - 64 a.a.) Partial Recombinant Protein

Product Code. 16231900
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16231900 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16231900 Supplier Abnova™ Supplier No. P3592.20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Sequence: VFGGIGDPVTCLKSGAICHPVFCPRRYKQIGTCGLPGTKCCKKP

Specifications

Accession Number O15263
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 100289462
Molecular Weight (g/mol) 4kDa
Name DEFB4B (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 20 μg
Immunogen VFGGIGDPVTCLKSGAICHPVFCPRRYKQIGTCGLPGTKCCKKP
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1 EU/μg)
Common Name DEFB4B
Gene Symbol NULL
Biological Activity The ED50 was determined by its ability to chemoattract immature human dendritic cells using a concentration range of 1-50ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.