missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dihydrofolate Reductase/DHFR Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7934
This item is not returnable.
View return policy
Description
Dihydrofolate Reductase/DHFR Polyclonal specifically detects Dihydrofolate Reductase/DHFR in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Dihydrofolate Reductase/DHFR | |
| Polyclonal | |
| Unconjugated | |
| P00374 | |
| DHFR | |
| E. coli-derived recombinant human Dihydrofolate Reductase/DHFR Met1-Asp187 Accession # P00374 | |
| 100 μg | |
| Primary | |
| Detects human Dihydrofolate Reductase/DHFR in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| DHFR, DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1719 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction