missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DIO2 (aa 50-123) Control Fragment Recombinant Protein

Product Code. 30201577
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
This item is not returnable. View return policy

Product Code. 30201577

missing translation for 'mfr': Invitrogen™ RP106163

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65489 (PA5-65489. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene belongs to the iodothyronine deiodinase family. It activates thyroid hormone by converting the prohormone thyroxine by outer ring deiodination to bioactive 3,3',5-triiodothyronine residues encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence, which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92813
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1734
Name Human DIO2 (aa 50-123) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5 DII; AI324267; D2; deiodinase, iodothyronine, type 2; deiodinase, iodothyronine, type II; deiodinase-2; deiodonase-2; Dio2; DIOII; iodothyronine type II deiodinase; Itdi2; SelY; thyroxine deiodinase; thyroxine deiodinase, type II; thyroxine type II; TXDI2; Type 2 DI; type 2 iodothyronine deiodinase; type II iodothyronine deiodinase; type-II 5' deiodinase; type-II 5'deiodinase; type-II 5'-deiodinase
Common Name DIO2
Gene Symbol DIO2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EWRRMLTSEGLRCVWKSFLLDAYKQVKLGEDAPNSSVVHVSSTEGGDNSGNGTQEKIAEGATCHLLDFASPERP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.