missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dkk-2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC6628U-100UG
This item is not returnable.
View return policy
Description
Dkk-2 Monoclonal specifically detects Dkk-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Dkk-2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| dickkopf (Xenopus laevis) homolog 2, dickkopf 2, dickkopf homolog 2 (Xenopus laevis), dickkopf related protein-2, Dickkopf-2, dickkopf-related protein 2, Dkk2, DKK-2, hDkk-2 | |
| Chinese Hamster Ovary cell line, CHO-derived human Dkk-2, Met1-Ile259, Accession # NP_055236 | |
| 100 μg | |
| 27123 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 994930 | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| NP_055236 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Dkk-2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction