missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dkk-4 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1269
This item is not returnable.
View return policy
Description
Dkk-4 Polyclonal specifically detects Dkk-4 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| Dkk-4 | |
| Polyclonal | |
| Biotin | |
| NP_055235 | |
| DKK4 | |
| Mouse myeloma cell line NS0-derived recombinant human Dkk-4 (R&D Systems, Catalog # 1269-DK) Leu19-Leu224 Accession # NP_055235 | |
| 50 μg | |
| Primary | |
| Detects human Dkk-4 in ELISAs and Western blots. In sandwich immunoassays, less than 2% cross-reactivity with recombinant mouse (rm) Dkk-4 is observed and less than 0.3% cross-reactivity with recombinant human (rh) Dkk-1, rmDkk-1, rhDkk-3, and recombinant rat Dkk-1 is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27121 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction