Learn More
Invitrogen™ Human DLX1 (aa 1-76) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-65154 (PA5-65154, PA5-83799 (PA5-83799. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | P56177 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 1745 |
| Name | Human DLX1 (aa 1-76) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | DII B; distal-less homeo box 1; distal-less homeobox 1; Dlx; Dlx1; Dlx-1; homeobox Dlx1; homeobox protein DLX-1 |
| Common Name | DLX1 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.