missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dopamine D1R/DRD1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8276U-100UG
This item is not returnable.
View return policy
Description
Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Dopamine D1R/DRD1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P21728 | |
| DRD1 | |
| NS0 mouse myeloma cell line transfected with human Dopamine D1 R/DRD1 Met1-Thr446 Accession # P21728 | |
| RUO | |
| 1812 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 887438 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 R, Dopamine D1 receptor, dopamine receptor D1, DRD1, DRD1A | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human Dopamine D1 R/DRD1 transfected cells but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction