missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPP6 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2360N-100UG
This item is not returnable.
View return policy
Description
DPP6 Monoclonal specifically detects DPP6 in Human samples. It is validated for Flow Cytometry.
Specifications
| DPP6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAA35761 | |
| DPP6 | |
| Mouse myeloma cell line NS0-derived recombinant human DPP6 Thr56-Asp803 (Glu458Val) Accession # AAA35761 | |
| RUO | |
| 1804 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 274308 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| dipeptidyl aminopeptidase IV-related protein, dipeptidyl aminopeptidase-like protein 6, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, dipeptidyl peptidase IV-related protein, Dipeptidyl peptidase VI, dipeptidylpeptidase 6, dipeptidyl-peptidase 6, dipeptidylpeptidase VI, DPP VI, DPPX, DPPXFLJ55680, MGC46605, VF2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human DPP6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DPPIV is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction