Learn More
Human Dynactin Subunit 1/DCTN1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand:  R&D Systems AF6657
Product Code. 18080612
  • € 581.00 / 100µg
Description
Dynactin Subunit 1/DCTN1 Polyclonal specifically detects Dynactin Subunit 1/DCTN1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dynactin Subunit 1/DCTN1
Polyclonal
Unconjugated
Q14203
DCTN1
E. coli-derived recombinant human Dynactin Subunit 1/DCTN1 Ala1145-Ser1278 Accession # Q14203
100 μg
Primary
Detects human Dynactin Subunit 1/DCTN1 in direct ELISAs and human, mouse and rat Dynactin Subunit 1/DCTN1 in Western blots.
Human
IgG
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
DAP-150, DCTN1, DP-150, Dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), HMN7B, p135, p150 Glued (Drosophila) homolog, p150-glued
Sheep
Affinity Purified
RUO
1639
Sterile PBS to a final concentration of 0.2 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.