Learn More
Human Dynactin Subunit 1/DCTN1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6657
This item is not returnable.
View return policy
Description
Dynactin Subunit 1/DCTN1 Polyclonal specifically detects Dynactin Subunit 1/DCTN1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Dynactin Subunit 1/DCTN1 | |
| Polyclonal | |
| Unconjugated | |
| Q14203 | |
| DCTN1 | |
| E. coli-derived recombinant human Dynactin Subunit 1/DCTN1 Ala1145-Ser1278 Accession # Q14203 | |
| 100 μg | |
| Primary | |
| Detects human Dynactin Subunit 1/DCTN1 in direct ELISAs and human, mouse and rat Dynactin Subunit 1/DCTN1 in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| DAP-150, DCTN1, DP-150, Dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), HMN7B, p135, p150 Glued (Drosophila) homolog, p150-glued | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1639 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |