missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EFCAB3 Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61356 (PA5-61356. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
EFCAB3 gene ontology annotations related to this gene include calcium ion binding.
Specifications
Specifications
| Accession Number | Q8N7B9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 146779 |
| Name | Human EFCAB3 Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | Efcab3; EF-hand calcium binding domain 3; EF-hand calcium-binding domain-containing protein 3; RGD1305423 |
| Common Name | EFCAB3 |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto