missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EGF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
€ 228.00 - € 2100.00
Specifications
| Antigen | EGF |
|---|---|
| Clone | 10825 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.05-0.1 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732757
![]() |
R&D Systems
MAB236-SP |
25 μg |
€ 228.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762759
![]() |
R&D Systems
MAB236-100 |
100 μg |
€ 519.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18784938
![]() |
R&D Systems
MAB236-500 |
500 μg |
€ 2100.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGF Monoclonal specifically detects EGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
| EGF | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.05-0.1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone | |
| EGF | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 10825 | |
| Western Blot, Immunohistochemistry, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 1950 | |
| E. coli-derived recombinant human EGF | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human EGF in direct ELISAs. Detects human EGF and rat EGF in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) HB-EGF or rhTGF-alpha is observed. In Western blots, no cross-reactivity with recombinant mouse EGF is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title