missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EGLN2/PHD1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF6394-SP
This item is not returnable.
View return policy
Description
EGLN2/PHD1 Polyclonal specifically detects EGLN2/PHD1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| EGLN2/PHD1 | |
| Polyclonal | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q96KS0 | |
| EGLN2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human EGLN2/PHD1 Asp2-Thr407 Accession # Q96KS0 | |
| 25 μg | |
| Primary | |
| Detects human EGLN2/PHD1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp434E026, EC 1.14.11, EC 1.14.11.-, egl nine homolog 2, egl nine homolog 2 (C. elegans), EIT6, Estrogen-induced tag 6, FLJ95603, HIFPH1, HIF-PH1, HIFPH1 EGL nine (C.elegans) homolog 2, HIF-prolyl hydroxylase 1, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 1, PHD1, Prolyl hydroxylase domain-containing protein 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 112398 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction