missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EMG1 (NP_006322, 1 a.a. - 244 a.a.) Full-length Recombinant Protein with His tag

Product Code. 16171870
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16171870 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16171870 Supplier Abnova™ Supplier No. P5850.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

EMG1 plays a role in the processing of the pre-18S rRNA and small ribosomal subunit assembly (Liu and Thiele, 2001 [PubMed 11694595]) and may play a role in methylation during ribosome biogenesis (Eschrich et al., 2002 [PubMed 11935223]).[supplied by OMIM]

Sequence: MGSSHHHHHHSSGLVPRGSHMGSMAAPSDGFKPRERSGGEQAQDWDALPPKRPRLGAGNKIGGRRLIVVLEGASLETVKVGKTYELLNCDKHKSILLKNGRDPGEARPDITHQSLLMLMDSPLNRAGLLQVYIHTQKNVLIEVNPQTRIPRTFDRFCGLMVQLLHKLSVRAADGPQKLLKVIKNPVSDHFPVGCMKVGTSFSIPVVSDVRELVPSSDPIVFVVGAFAHGKVSVEYTEKMVSISNYPLSAALTCAKLTTAFEEVWGVI

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 10436
Molecular Weight (g/mol) 29.1kDa
Name EMG1 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 3 ug/lane in 15% SDS-PAGE Stained with Coomassie Blue. Due to the protein nature, dimmers and multimers may be observed.
Quantity 100 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMGSMAAPSDGFKPRERSGGEQAQDWDALPPKRPRLGAGNKIGGRRLIVVLEGASLETVKVGKTYELLNCDKHKSILLKNGRDPGEARPDITHQSLLMLMDSPLNRAGLLQVYIHTQKNVLIEVNPQTRIPRTFDRFCGLMVQLLHKLSVRAADGPQKLLKVIKNPVSDHFPVGCMKVGTSFSIPVVSDVRELVPSSDPIVFVVGAFAHGKVSVEYTEKMVSISNYPLSAALTCAKLTTAFEEVWGVI
Storage Requirements Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C2F/FLJ60792/Grcc2f/NEP1
Common Name EMG1
Gene Symbol EMG1
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS - PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.