missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human EMMPRIN/CD147 Quantikine ELISA Kit

Product Code. 18762074
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18762074

Brand: R&D Systems™ DEMP00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human EMMPRIN/CD147 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human EMMPRIN/CD147 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human EMMPRIN/CD147 in cell culture supernates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (50 ul), urine (10 ul), human milk (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 62.50 to 4000 pg/mL
Assay Sensitivity 9.77 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human EMMPRIN
For Use With (Application) ELISA
Gene ID (Entrez) 682
Gene Symbol BSG
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Target EMMPRIN/CD147
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.