missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EMP3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4505G-100UG
This item is not returnable.
View return policy
Description
EMP3 Monoclonal specifically detects EMP3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| EMP3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P54852 | |
| EMP3 | |
| NS0 mouse myeloma cell line transfected with human EMP3 Met1-Glu163 Accession # P54852 | |
| RUO | |
| 2014 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 439949 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EMP-3, epithelial membrane protein 3, HNMP-1, Protein YMP, YMP, YMPHematopoietic neural membrane protein 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human EMP. Stains human EMP3 cell transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction