missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EMR3 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8496R-100UG
This item is not returnable.
View return policy
Description
EMR3 Monoclonal specifically detects EMR3 in Human samples. It is validated for Flow Cytometry.
Specifications
| EMR3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BY15 | |
| EMR3 | |
| NS0 mouse myeloma cell line transfected with human EMR3 Met1-Tyr652 Accession # Q9BY15 | |
| RUO | |
| 84658 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 908235 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| egf-like module containing, mucin-like, hormone receptor-like 3, EGF-like module receptor 3, EGF-like module-containing mucin-like hormone receptor-like 3, egf-like module-containing mucin-like receptor 3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human EMR3 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction