missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Endorepellin/Perlecan Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2364
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Polyclonal specifically detects Endorepellin/Perlecan/Heparan Sulfate Proteoglycan in Human samples. It is validated for Western Blot.
Specifikationer
| Endorepellin/Perlecan/Heparan Sulfate Proteoglycan | |
| Polyclonal | |
| Biotin | |
| P98160 | |
| HSPG2 | |
| Mouse myeloma cell line NS0-derived recombinant human Endorepellin/Perlecan Arg3684-Ser4391 (Ser4003Thr) Accession # P98160 | |
| 50 μg | |
| Primary | |
| Detects human Endorepellin/Perlecan in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3339 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering