missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ENPP-1 APC-conjugated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems FAB6136A
This item is not returnable.
View return policy
Description
ENPP-1 Polyclonal specifically detects ENPP-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| ENPP-1 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, ENPP1, Ly-41 Antigen, M6S1, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPP1, NPPS, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1, PCA1ARHR2, PDNP1, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5167 | |
| Human | |
| IgG |
| Flow Cytometry | |
| APC | |
| Q13822 (ENPP-2) P22413 (ENPP-1) | |
| ENPP1 | |
| Mouse myeloma cell line NS0-derived recombinant human ENPP-2-1-2 Asp49-Trp144 (ENPP-2), Val191-Leu591 (ENPP-1), Asn532-Ile863 (ENPP-2). Cross-reactivity with human ENPP-2 was removed from the final product. Accession # Q13822 (ENPP-2) P22413 (ENPP-1) | |
| 100 Tests | |
| Primary | |
| Detects human ENPP-1 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human ENPP-2 and recombinant mouse ENPP-2 is observed after removal of cross-reactivity with human ENPP-2. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction