missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EOMES Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC6166V-100UG
This item is not returnable.
View return policy
Description
EOMES Monoclonal specifically detects EOMES in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| EOMES | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O95936 | |
| EOMES | |
| E. coli-derived recombinant human EOMES Gly471-Pro686 Accession # O95936 | |
| RUO | |
| 8320 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 644730 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR2, TBR-2, TBR2eomesodermin homolog, T-brain-2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human EOMES in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Brachyury, rhEOMES (aa 1-115), recombinant mouse EOMES (aa 1-126), rhTBX2, 3, 5, 6, 18, or 20 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction