missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCL11 (Eotaxin) Recombinant Protein
Click to view available options
Quantity:
10 μg
100 μg
25 μg
Unit Size:
100µg
10µg
25µg
Description
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications
Specifications
| Accession Number | P51671 |
| Concentration | 200 μg/mL |
| For Use With (Application) | Control, ELISA, Western Blot |
| Formulation | PBS with no preservative |
| Gene ID (Entrez) | 6356 |
| Molecular Weight (g/mol) | 8.4 kD |
| Name | Human Eotaxin (aa 24-97) |
| Quantity | 25 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction