missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EphB6 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB33841N-100UG
This item is not returnable.
View return policy
Description
EphB6 Monoclonal specifically detects EphB6 in Human samples. It is validated for Flow Cytometry.
Specifications
| EphB6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O15197 | |
| EPHB6 | |
| Mouse myeloma cell line NS0-derived recombinant human EphB6 Val17-Thr579 Accession # O15197 | |
| RUO | |
| 2051 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 465327 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, Hep, HEPTyrosine-protein kinase-defective receptor EPH-6, Mep, MGC129910, MGC129911 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human EphB6 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction