missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ephrin-A5 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF3743
This item is not returnable.
View return policy
Description
Ephrin-A5 Polyclonal specifically detects Ephrin-A5 in Human samples. It is validated for Western Blot.
Specifications
| Ephrin-A5 | |
| Polyclonal | |
| Biotin | |
| P52803 | |
| EFNA5 | |
| Mouse myeloma cell line NS0-derived recombinant human Ephrin-A5 Gln21-Asn203 Accession # P52803 | |
| 50 μg | |
| Primary | |
| Detects human Ephrin-A5 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Ephrin-A2 and rmEphrin-B2 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AF1, AL-1, EFL-5, EFNA5, EPH-related receptor tyrosine kinase ligand 7, ephrin-A5, EphrinA5, EPLG7, GLC1M, LERK-7, LERK7EFL5, RAGS | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1946 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction