Learn More
Human Epithelial Cell Mouse, anti-Human, Unconjugated, Clone: 5E11.3.1, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60147
This item is not returnable.
View return policy
Description
- The 5E11.3.1 antibody reacts with an unidentified marker expressed on the surface of most luminal cells of normal human mammary epithelium
- Weak labeling (or no labeling) is observed within the basal cell compartment of the mammary epithelium
- The 5E11.3.1 antibody gives variable labeling on malignant breast epithelium
- Positive labeling is observed on the following cell lines: MCF7, CAMA, T47D, BT-10, BT-20, COLO 205, and WiDr
- Labeling is negative on human bone marrow cells, blood cells, lymphoid and mesenchymal tissues, mammary and marrow fibroblast cultures, and human embryonic stem cell line H9
Specifications
| Human Epithelial Cell | |
| Monoclonal | |
| Unconjugated | |
| 100 μg | |
| Human | |
| Mammary Cells |
| Immunohistochemistry, Flow Cytometry | |
| 5E11.3.1 | |
| Mouse | |
| Primary | |
| IgG1 κ |