Learn More
Human Epithelial Cell Mouse, anti-Human, Unconjugated, Clone: 5E11.3.1, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60147
Encompass_Preferred
Product Code. 17179181
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • The 5E11.3.1 antibody reacts with an unidentified marker expressed on the surface of most luminal cells of normal human mammary epithelium
  • Weak labeling (or no labeling) is observed within the basal cell compartment of the mammary epithelium
  • The 5E11.3.1 antibody gives variable labeling on malignant breast epithelium
  • Positive labeling is observed on the following cell lines: MCF7, CAMA, T47D, BT-10, BT-20, COLO 205, and WiDr
  • Labeling is negative on human bone marrow cells, blood cells, lymphoid and mesenchymal tissues, mammary and marrow fibroblast cultures, and human embryonic stem cell line H9
Specifications
Specifications
Human Epithelial Cell
Monoclonal
Unconjugated
100 μg
Human
Mammary Cells
Immunohistochemistry, Flow Cytometry
5E11.3.1