missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Eps15 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC8480G-100UG
This item is not returnable.
View return policy
Description
EPS15 Monoclonal specifically detects EPS15 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| EPS15 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15 | |
| E.coli-derived recombinant human Eps15, Ala448-Thr579, Accession # P42566 | |
| 100 μg | |
| Primary | |
| Detects human Eps15 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
| Flow Cytometry | |
| 1261C | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P42566 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2060 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction