Learn More
R&D Systems™ Human ErbB2/Her2 (NP_004439) VersaClone cDNA
Product Code. 18758388
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 18758388
Brand: R&D Systems™ RDC1022
v-erb-b2 avian erythroblastic leukemia viral oncogene homolog
The Human ErbB2/Her2 (NP_004439) VersaClone cDNA clone contains the complete ORF for ErbB2/Her2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ErbB2/Her2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans