missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ERMAP Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB49281S-100UG
This item is not returnable.
View return policy
Description
ERMAP Monoclonal specifically detects ERMAP in Human samples. It is validated for Flow Cytometry.
Specifications
| ERMAP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96PL5 | |
| ERMAP | |
| Mouse myeloma cell line NS0-derived recombinant human ERMAP His30-Ala155 Accession # Q96PL5 | |
| RUO | |
| 114625 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 767505 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| erythroblast membrane-associated protein, erythroblast membrane-associated protein (RD and SC blood groups), erythroblast membrane-associated protein (Scianna blood group), erythroid membrane-associated protein, hERMAP, MGC118810, MGC118811, PRO2801, Radin blood group, Radin blood group (Rd), Radin blood group antigen, RD, RDMGC118812, SC, Scianna blood group, Scianna blood group (Sc), Scianna blood group antigen, SCMGC118813 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ERMAP in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction