missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin R Fluorescein-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB307F
This item is not returnable.
View return policy
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Flow Cytometry.
Specifications
| Erythropoietin R | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2057 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 38409 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P19235 | |
| EPOR | |
| Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Pro26-Pro250 Accession # P19235 | |
| 100 Tests | |
| Primary | |
| Detects human Erythropoietin R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Erythropoietin R is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction