missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FABP3/H-FABP Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF1678
This item is not returnable.
View return policy
Description
FABP3/H-FABP Polyclonal specifically detects FABP3/H-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FABP3/H-FABP | |
| Polyclonal | |
| Biotin | |
| P05413 | |
| FABP3 | |
| E. coli-derived recombinant human FABP3 Met1-Ala133 Accession # P05413 | |
| 50 μg | |
| Primary | |
| Detects human FABP3 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse (rm) FABP4 is observed, 20% cross-reactivity with recombinant rat (rr) FABP1 is observed, and 10% cross-reactivity with rrFABP2 and rmFABP5 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, HFABP, H-FABP, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2170 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction