missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FABP6 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3880-SP
This item is not returnable.
View return policy
Description
FABP6 Polyclonal specifically detects FABP6 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| FABP6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| fatty acid binding protein 6, ileal, Fatty acid-binding protein 6, gastrotropin, I-15P, I-15PIntestinal 15 kDa protein, IBABP, I-BABPIntestinal bile acid-binding protein, I-BALB, I-BAP, ILBP, ILBP3, ILBPGT, ileal bile acid binding protein, ILLBP, ILLBPIleal lipid-binding protein, illeal lipid-binding protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2172 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P51161 | |
| FABP6 | |
| E. coli-derived recombinant human FABP6 Ala2-Ala128 Accession # P51161 | |
| 25 μg | |
| Primary | |
| Detects human FABP6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) FABP1, rhFABP2, rhFABP3, rhFABP4, rhFABP5, rhFABP7, and recombinant mouse FABP9 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction