Learn More
Invitrogen™ Human FAM132A (aa 70-138) Control Fragment Recombinant Protein
Product Code. 30204407
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30204407 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30204407 Supplier Invitrogen™ Supplier No. RP90538
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54980 (PA5-54980. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Insulin-sensitizing adipocyte-secreted protein (adipokine) that regulates glucose metabolism in liver and adipose tissue. Promotes glucose uptake in adipocytes and suppresses de novo glucose production in hepatocytes via the PI3K-Akt signaling pathway. Administration lead to reduction of blood glucose. Able to attenuate inflammation in fat tissue. [UniProt]
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q5T7M4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 388581
Name Human FAM132A (aa 70-138) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Adipolin; Adipolin cleaved form; Adipolin fC1QTNF12; Adipolin fCTRP12; Adipolin full-length form; Adipolin gC1QTNF12; Adipolin gCTRP12; Adipose-derived insulin-sensitizing factor; C1q and TNF related 12; C1q and TNF related protein 12; C1q domain containing 2; C1q/TNF-related protein 12; C1QDC2; C1QTNF12; Complement C1q tumor necrosis factor-related protein 12; CTRP12; FAM132A; family with sequence similarity 132 member A; family with sequence similarity 132, member A; protein FAM132A; zgc:158358
Common Name FAM132A
Gene Symbol C1QTNF12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DAHMTWLNFVRRPDDGALRKRCGSRDKKPRDLFGPPGPPGAEVTAETLLHEFQELLKEATERRFSGLLD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining