missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FAM3C Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF3084
This item is not returnable.
View return policy
Description
FAM3C Polyclonal specifically detects FAM3C in Human samples. It is validated for Western Blot.
Specifications
| FAM3C | |
| Polyclonal | |
| Biotin | |
| Q92520 | |
| FAM3C | |
| Mouse myeloma cell line NS0-derived recombinant human FAM3C Gln25-Asp227 Accession # Q92520 | |
| 50 μg | |
| Primary | |
| Detects human FAM3C in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse FAM3C is observed and 5% cross-reactivity with recombinant human (rh) FAM3A, rhFAM3B, and rhFAM3D is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| D6WSU176e, family with sequence similarity 3, member C, GS3876, ILEI, ILEIInterleukin-like EMT inducer, predicted osteoblast protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10447 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction