missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human FAP Protein, Fc Tag (active enzyme)

Product Code. 17146291
Change view
Click to view available options
Quantity:
100 μg
20 μg
500 μg
Unit Size:
100µg
20µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17146291 100 μg 100µg
17166291 500 μg 500µg
17156291 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17146291 Supplier ACROBiosystems Supplier No. FAPH5263100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FAP (also known as seprase) is a Type II transmembrane serine protease. Both plasma membrane and soluble forms exhibit post-proline cleaving endopeptidase activity, with a marked preference for Ala/Ser-Gly-Pro-Ser/Asn/Ala consensus sequences. Degrade also gelatin, heat-denatured type I collagen. Also has dipeptidyl peptidase activity, with a preference for Ala-Pro, Ile-Pro, Gly-Pro, Arg-Pro and Pro-Pro. The plasma membrane form, in association with either DPP4, PLAUR or integrins, is involved in the pericellular proteolysis of the extracellular matrix (ECM), and hence promotes cell adhesion, migration and invasion through the ECM. Promotes glioma cell invasion through the brain parenchyma by degrading the proteoglycan brevican. Acts as a tumor suppressor in melanocytic cells through regulation of cell proliferation and survival in a serine protease activity-independent manner.

This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 111.5 kDa. The protein migrates as 110-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Molecular Weight (g/mol) 111.5 kDa
Quantity 100 μg
Source Human FAP, Fc Tag (FAP-H5263) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # Q12884-1).
Immunogen Leu 26 - Asp 760
Storage Requirements -20°C
Gene Alias FAP,FAPalpha,SIMP,Seprase,APCE
Common Name FAP
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Enzymes
Purity or Quality Grade >90% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.