missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fas Ligand/TNFSF6 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF126-SP
This item is not returnable.
View return policy
Description
Fas Ligand/TNFSF6 Polyclonal specifically detects Fas Ligand/TNFSF6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization.
Specifications
| Fas Ligand/TNFSF6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 356 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.012-0.072 ug/mL | |
| Q53ZZ1 | |
| FASLG | |
| Chinese hamster ovary cell line CHO-derived recombinant human Fas Ligand/TNFSF6 Pro134-Leu281 Accession # Q53ZZ1 | |
| 25 μg | |
| Primary | |
| Detects human Fas Ligand in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Fas Ligand is observed, approximately 7% cross-reactivity with recombinant rat Fas Ligand and recombinant human (rh) BAFF, and less than 1% cross-reactivity with rhTRAIL, rhTNF-alpha , rhGITR Ligand, and rhAPRIL is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction