missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RI/CD64 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB12571R-100UG
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry.
Specifications
| Fc gamma RI/CD64 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P12314.2 | |
| FCGR1A | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RI Gln16-Pro288 Accession # P12314.2 | |
| RUO | |
| 2209 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 276426 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Fc gamma RI in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) Fc gamma RI, rmFc gamma RII, recombinant human (rh) Fc gamma RIIA, rhFc gamma RIIB, or rhFc gamma RIIIB. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction