missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Fc gamma RIIA / CD32a (H167) Protein, His Tag

Product Code. 15806147
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15806147 1 mg 1mg
15896137 100 μg 100µg
17176111 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15806147 Supplier ACROBiosystems Supplier No. CD1H52231MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SPR & BLI verified

Receptors for the Fc region of IgG (Fc γ R) are members of the Ig superfamily that function in the activation or inhibition of immune responses. Three classes of human Fc γ Rs: RI (CD64), RII (CD32), and RIII (CD16), which generate multiple isoforms, are recognized. There are three genes for human Fcγ RII /CD32 (A, B, and C) and one for mouse Fcγ RII B (CD32B). CD32 is a low affinity receptor for IgG. The activating isoform, CD32A, is expressed on monocytes, neutrophils, platelets and dendritic cells. CD32A is expressed on many immune cell types (macrophage, neutrophil, eosinophils, platelets, dendritic cells and Langerhan cells), where inhibitory ITIM­bearing receptors may also be coexpressed and co­engaged by specific ligands. CD32A delivers an activating signal upon ligand binding, and results in the initiation of inflammatory responses including cytolysis, phagocytosis, degranulation and cytokine production. The responses can be modulated by signals from the coexpressed inhibitory receptors such as CD32B, and the strength of the signal is dependent on the ratio of expression of the activating and inhibitory receptors.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.5 kDa. The protein migrates as 29-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number AAH19931.1
Molecular Weight (g/mol) 21.5 kDa
Quantity 1 mg
Source Human CD32a (H167), His Tag (SPR and BLI verified) (CD1-H5223) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1).
Immunogen Ala 36 - Ile 218
Storage Requirements -20°C
Gene Alias CD32a,FCGR2A,CD32,FCG2 ,FCGR2A1,IGFR2
Common Name Fc gamma RIIA / CD32a
Conjugate Unconjugated
Species Human
Protein Tag His Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype MABSol™ Fc Receptors
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.