missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRL1/FcRH1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2049T-100UG
This item is not returnable.
View return policy
Description
FCRL1/FcRH1 Monoclonal specifically detects FCRL1/FcRH1 in Human samples. It is validated for Flow Cytometry.
Specifications
| FCRL1/FcRH1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96LA6 | |
| FCRL1 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL1/FcRH1 Ala17-His304 Accession # Q96LA6 | |
| RUO | |
| 115350 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 282415 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| BXMH1, CD307a, CD307a antigen, Fc receptor homolog 1, Fc receptor-like 1, FcRH1, FCRH1DKFZp667O1421, FcRL1, FcR-like protein 1, hIFGP1, IFGP family protein 1, IFGP1, IFGP1Fc receptor-like protein 1, Immune receptor translocation-associated protein 5, IRTA5, IRTA5immunoglobulin superfamily Fc receptor, gp42 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FCRL1/FcRH1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) FCRL2, 3, 4, or 5 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction