missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRL4/FcRH4 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB24262S-100UG
This item is not returnable.
View return policy
Description
FCRL4/FcRH4/IRTA1 Monoclonal specifically detects FCRL4/FcRH4/IRTA1 in Human samples. It is validated for Flow Cytometry.
Specifications
| FCRL4/FcRH4/IRTA1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_112572 | |
| FCRL4 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL4/FcRH4 Gln16-Asp385 Accession # NP_112572 | |
| RUO | |
| 83417 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 580810 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD307d, CD307d antigen, Fc receptor homolog 4, Fc receptor-like 4, FcRH4, FCRH4Fc receptor-like protein 4, FcRL4, FcR-like protein 4, hIFGP2, IFGP family protein 2, IFGP2, IGFP2, Immune receptor translocation-associated protein 1, immunoglobulin superfamily Fc receptor, gp42, immunoglobulin superfamily receptor translocation associated 1, IRTA1, IRTA1MGC150523, MGC150522 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FCRL4/FcRH4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human FCRL1 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction