missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ferritin Light Chain Mouse, Clone: FTL-Q2, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QDX15-100ug
This item is not returnable.
View return policy
Specifications
| Human Ferritin Light Chain | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 100 μg | |
| Primary | |
| IgG1 |
| ELISA, Lateral Flow | |
| FTL-Q2 | |
| PBS (pH 7.4) | |
| Protein G | |
| RUO | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction