missing translation for 'onlineSavingsMsg'
Learn More

Human FGF-18 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

€ 234.00 - € 536.00

Specifications

Antigen FGF18
Clone 91811
Dilution Immunohistochemistry 5-15 μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18714514
GREENER_CHOICE
View Documents
R&D Systems
MAB6671-100
100 μg
€ 536.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18794544
View Documents
R&D Systems
MAB6671-SP
25 μg
€ 234.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FGF18 Monoclonal antibody specifically detects FGF18 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

FGF18
Immunohistochemistry 5-15 μg/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
FGF18, FGF-18ZFGF5, fibroblast growth factor 18, zFGF5
E. coli-derived human FGF-18 protein, Glu28-Ala207, Accession # O76093
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
91811
Immunohistochemistry
Unconjugated
Mouse
Human
O76093
8817
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.