missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-21, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17826518
Click to view available options
Quantity:
250 μg
5 μg
Unit Size:
250µg
5µg
This item is not returnable. View return policy

Product Code. 17826518

Brand: Gibco™ AF100425UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human FGF-21 is a 19.5 kDa protein containing 182 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NSA1
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 26291
Molecular Weight (g/mol) 19.5 kDa
Name Human FGF-21, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 5 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196
Common Name FGF21
Gene Symbol FGF21
Biological Activity Determined by its ability to stimulate the proliferation of murine NIH-3T3 cells. The expected ED50 for this effect is 0.3-0.6 ug/ml, in the presence of murine Klotho.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of murine NIH-3T3 cells. The expected ED50 for this effect is 0.3-0.6 ug/ml, in the presence of murine Klotho.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.