Learn More
Mouse Axl Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 17 publications
Brand: R&D Systems AF854-SP
This item is not returnable.
View return policy
Description
Axl Polyclonal specifically detects Axl in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Axl | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 558 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q80YQ3 | |
| AXL | |
| Mouse myeloma cell line NS0-derived recombinant mouse Axl His20-Pro443 Accession # Q80YQ3 | |
| 25 μg | |
| Primary | |
| Detects mouse Axl in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human Axl is observed. | |
| Mouse | |
| IgG |