missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FGF22 (Q9HCT0) Recombinant Protein

Product Code. 16131530
Click to view available options
:
20μg
This item is not returnable. View return policy

Product Code. 16131530

Brand: Abnova™ P4383.20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The mouse homolog of this gene was found to be preferentially expressed in the inner root sheath of the hair follicle, which suggested a role in hair development. [provided by RefSeq]

Sequence: MTPSASRGPRSYPHLEGDVRWRRLFSSTHFFLRVDPGGRVQGTRWRHGQDSILEIRSVHVGVVVIKAVSSGFYVAMNRRGRLYGSRLYTVDCRFRERIEENGHNTYASQRWRRRGQPMFLALDRRGGPRPGGRTRRYHLSAHFLPVLVS

Specifications

Accession Number Q9HCT0
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 27006
Molecular Weight (g/mol) 17.3kDa
Name FGF22 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
Quantity 20 ug
Immunogen MTPSASRGPRSYPHLEGDVRWRRLFSSTHFFLRVDPGGRVQGTRWRHGQDSILEIRSVHVGVVVIKAVSSGFYVAMNRRGRLYGSRLYTVDCRFRERIEENGHNTYASQRWRRRGQPMFLALDRRGGPRPGGRTRRYHLSAHFLPVLVS
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Common Name FGF22
Gene Symbol FGF22
Biological Activity Activity was determined by the dose-dependent proliferation of 4MBr-5 cells, is typically 50-300ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.