missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGFR3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB766G-100UG
This item is not returnable.
View return policy
Description
FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Flow Cytometry.
Specifications
| FGFR3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2261 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 136334 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FGFR3 | |
| Pool of NS0-derived recombinant human FGF R3 alpha (IIIb) and Sf21-derived FGF R3 alpha (IIIc) | |
| 100 μg | |
| Primary | |
| Detects the IIIb and IIIc isoforms of human FGF R3 in direct ELISAs and Western blots. Does not cross-react with any isoforms of recombinant mouse (rm) FGF R3, rmFGF R2, recombinant human (rh) FGF R1, rhFGF R2, or rhFGF R4. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction