Learn More
Human Fibronectin Fluorescein-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems IC1918F
Product Code. 18173731
  • € 415.00 / 100 tests
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibronectin
Monoclonal
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
FN1
Human plasma-derived Fibronectin
100 Tests
Primary
Detects human Fibronectin in Western blots.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
P1H11
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2335