missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FKBP2 (NP_001128680, 22 a.a. - 142 a.a.) Partial Recombinant Protein

Product Code. 16151400
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 16151400

Brand: Abnova™ P3503.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It is thought to function as an ER chaperone and may also act as a component of membrane cytoskeletal scaffolds. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MATGAEGKRKLQIGVKKRVDHCPIKSRKGDVLHMHYTGKLEDGTEFDSSLPQNQPFVFSLGTGQVIKGWDQGLLGMCEGEKRKLVIPSELGYGERGAPPKIPGGATLVFEVELLKIERRTEL

Specifications

Accession Number NP_001128680
Concentration 1 mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 2286
Molecular Weight (g/mol) 13.4kDa
Name FKBP2 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS-PAGE
Quantity 50 μg
Immunogen MATGAEGKRKLQIGVKKRVDHCPIKSRKGDVLHMHYTGKLEDGTEFDSSLPQNQPFVFSLGTGQVIKGWDQGLLGMCEGEKRKLVIPSELGYGERGAPPKIPGGATLVFEVELLKIERRTEL
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FKBP-13/PPIase
Common Name FKBP2
Gene Symbol FKBP2
Biological Activity Specific activity is > 270nmoles/min/μg, and is defined as the amount of enzyme that cleaves 1μmole of suc-AAPF-pNA per minute at 25°C in Tris-HCl pH 8.0 using chymotrypsin.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.