missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Flt-3 Ligand/FLT3L Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems BAF308
This item is not returnable.
View return policy
Description
Flt-3 Ligand/FLT3L Polyclonal specifically detects Flt-3 Ligand/FLT3L in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| Flt-3 Ligand | |
| Polyclonal | |
| Biotin | |
| AAA17999 | |
| FLT3LG | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Flt-3 Ligand (R&D Systems, Catalog # 308-FK) Thr27-Pro185 Accession # AAA17999 | |
| 50 μg | |
| Primary | |
| Detects human Flt-3 ligand in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with rhPlGF, rmFlt-3 Ligand, rhVEGF and rhFlt-1 Fc chimera is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2323 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction