missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FOXA2 (aa 391-443) Control Fragment Recombinant Protein

Product Code. 30206438
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206438

Brand: Invitrogen™ RP107525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y261
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3170
Name Human FOXA2 (aa 391-443) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3 beta; HNF3B; Hnf-3 b; HNF3beta; HNF-3 BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3 B; transcription factor; transcription factor 3 B; transcription factor Foxa2; WHN
Common Name FOXA2
Gene Symbol FOXA2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SHHHHQPHKMDLKAYEQVMHYPGYGSPMPGSLAMGPVTNKTGLDASPLAADTS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.