missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FoxD3 aa 1-140 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF2819-SP
This item is not returnable.
View return policy
Description
FoxD3 Polyclonal specifically detects FoxD3 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| FoxD3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AIS1, CWH3, forkhead box D3, forkhead box protein D3, Genesis, HFH2, HNF3/FH transcription factor genesis | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 27022 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q9UJU5 | |
| FOXD3 | |
| E. coli-derived recombinant human FoxD3 Met1-Val140 Accession # Q9UJU5 | |
| 25 μg | |
| Primary | |
| Detects human FoxD3 (aa 1-140) in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FoxP3, rhFoxP1, and rhFoxO3A is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction